Skip to Content
Merck
N-Nitroso Methylphenidate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

N-Nitroso Methylphenidate

N-Nitroso Methylphenidate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Methyl 2-(1-nitrosopiperidin-2-yl)-2-phenylacetate, Methyl-1-nitroso-α-phenyl-2-piperidineacetate
Empirical Formula (Hill Notation):
C14H18N2O3
CAS Number:
Molecular Weight:
262.30

Products

Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Technique