Skip to Content
Merck
Piperazine Dihydrochloride (Piperazine Dihydrochloride Hydrate) certified reference material, pharmaceutical secondary standard

Piperazine Dihydrochloride (Piperazine Dihydrochloride Hydrate)

Piperazine Dihydrochloride (Piperazine Dihydrochloride Hydrate) certified reference material, pharmaceutical secondary standard
Synonym(s):
Piperazine dihydrochloride
Empirical Formula (Hill Notation):
C4H10N2 · 2HCl · xH2O
Molecular Weight:
159.06 (anhydrous basis)

Products

Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade