Skip to Content
Merck
Potassium Clavulanate EP Impurity D certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Potassium Clavulanate EP Impurity D

Potassium Clavulanate EP Impurity D certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
4-(2-Hydroxyethyl)-1H-pyrrole-3-carboxylic acid
Empirical Formula (Hill Notation):
C7H9NO3
CAS Number:
Molecular Weight:
155.15

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Reference Material Type