
Rapamycin from Streptomyces hygroscopicus

Synonym(s):
Rapamycin from Streptomyces hygroscopicus, 23,27-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine, AY 22989, Sirolimus
Empirical Formula (Hill Notation):
C51H79NO13
CAS Number:
Molecular Weight:
914.17
Products
Product Category
Brand
Formula Weight
Gene Alias
Product Line
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing