
Tioconazole Related Compound B

Synonym(s):
1-[2,4-Dichloro-β-[(2,5-dichloro-3-thenyl)oxy]phenethyl]imidazole hydrochloride hydrochloride
Empirical Formula (Hill Notation):
C16H14Cl4N2OS · HCl
CAS Number:
Molecular Weight:
460.63
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2