Skip to Content
Merck
TOYOPEARL® SuperButyl-550C Bulk Media bottle of 1 L, phase C4 (butyl), 100 μm particle size

TOYOPEARL® SuperButyl-550C Bulk Media

TOYOPEARL® SuperButyl-550C Bulk Media bottle of 1 L, phase C4 (butyl), 100 μm particle size
Synonym(s):
TOYOPEARL® SuperButyl-550 HIC Bulk Media

Products

Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique