
Triclocarban Related Compound A

Synonym(s):
1,3-Bis(4-chlorophenyl)urea, 4,4′-Dichlorocarbanilide
Empirical Formula (Hill Notation):
C13H10Cl2N2O
CAS Number:
Molecular Weight:
281.14
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2