Skip to Content
Merck
Ytterbium(III) oxide 99.99% trace metals basis

Ytterbium oxide

Ytterbium(III) oxide 99.99% trace metals basis
Linear Formula:
Yb2O3
CAS Number:
Molecular Weight:
394.08
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing