Merck
  • First-in-Human Phase I Study of the Tamoxifen Metabolite Z-Endoxifen in Women With Endocrine-Refractory Metastatic Breast Cancer.
First-in-Human Phase I Study of the Tamoxifen Metabolite Z-Endoxifen in Women With Endocrine-Refractory Metastatic Breast Cancer.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2017-08-31)
Matthew P Goetz, Vera J Suman, Joel M Reid, Don W Northfelt, Michael A Mahr, Andrew T Ralya, Mary Kuffel, Sarah A Buhrow, Stephanie L Safgren, Renee M McGovern, John Black, Travis Dockter, Tufia Haddad, Charles Erlichman, Alex A Adjei, Dan Visscher, Zachary R Chalmers, Garrett Frampton, Benjamin R Kipp, Minetta C Liu, John R Hawse, James H Doroshow, Jerry M Collins, Howard Streicher, Matthew M Ames, James N Ingle
ABSTRACT
Purpose Endoxifen is a tamoxifen metabolite with potent antiestrogenic activity. Patients and Methods We performed a phase I study of oral Z-endoxifen to determine its toxicities, maximum tolerated dose (MTD), pharmacokinetics, and clinical activity. Eligibility included endocrine-refractory, estrogen receptor-positive metastatic breast cancer. An accelerated titration schedule was applied until moderate or dose-limiting toxicity occurred, followed by a 3+3 design and expansion at 40, 80, and 100 mg per day. Tumor DNA from serum (circulating cell free [cf); all patients] and biopsies [160 mg/day and expansion]) was sequenced. Results Of 41 enrolled patients, 38 were evaluable for MTD determination. Prior endocrine regimens during which progression occurred included aromatase inhibitor (n = 36), fulvestrant (n = 21), and tamoxifen (n = 15). Patients received endoxifen once daily at seven dose levels (20 to 160 mg). Dose escalation ceased at 160 mg per day given lack of MTD and endoxifen concentrations > 1,900 ng/mL. Endoxifen clearance was unaffected by CYP2D6 genotype. One patient (60 mg) had cycle 1 dose-limiting toxicity (pulmonary embolus). Overall clinical benefit rate (stable > 6 months [n = 7] or partial response by RECIST criteria [n = 3]) was 26.3% (95% CI, 13.4% to 43.1%) including prior tamoxifen progression (n = 3). cfDNA mutations were observed in 13 patients ( PIK3CA [n = 8], ESR1 [n = 5], TP53 [n = 4], and AKT [n = 1]) with shorter progression-free survival ( v those without cfDNA mutations; median, 61 v 132 days; log-rank P = .046). Clinical benefit was observed in those with ESR1 amplification (tumor; 80 mg/day) and ESR1 mutation (cfDNA; 160 mg/day). Comparing tumor biopsies and cfDNA, some mutations ( PIK3CA, TP53, and AKT) were undetected by cfDNA, whereas cfDNA mutations ( ESR1, TP53, and AKT) were undetected by biopsy. Conclusion In endocrine-refractory metastatic breast cancer, Z-endoxifen provides substantial drug exposure unaffected by CYP2D6 metabolism, acceptable toxicity, and promising antitumor activity.
Sigma-Aldrich
(Z)-Endoxifen, ≥98% (HPLC)