Merck
  • Characterization of Zaire ebolavirus glycoprotein-specific monoclonal antibodies.
Characterization of Zaire ebolavirus glycoprotein-specific monoclonal antibodies.
Clinical immunology (Orlando, Fla.) (2011-09-20)
Xiangguo Qiu, Judie B Alimonti, P Leno Melito, Lisa Fernando, Ute Ströher, Steven M Jones
ABSTRACT
Zaire ebolavirus (ZEBOV) can be transmitted by human-to-human contact and causes acute haemorrhagic fever with case fatality rates up to 90%. There are no effective therapeutic or prophylactic treatments available. The sole transmembrane glycoprotein (GP) is the key target for developing neutralizing antibodies. In this study, recombinant VSVΔG/ZEBOVGP was used to generate monoclonal antibodies (MAbs) against the ZEBOV GP. A total of 8 MAbs were produced using traditional hybridoma cell fusion technology, and then characterized by ELISA using ZEBOV VLPs, Western blotting, an immunofluorescence assay, and immunoprecipitation. All 8 MAbs worked in IFA and IP, suggesting that they are all conformational MAbs, however six of them recognized linearized epitopes by WB. ELISA results demonstrated that one MAb bound to a secreted GP (sGP 1-295aa); three bind to a part of the mucin domain (333-458aa); three MAbs recognized epitopes on the C-terminal domain of GP1 (296-501aa); and one bound to full length GP (VLPs/GP1,2 ΔTm). Using a mouse model these MAbs were evaluated for their therapeutic capacity during a lethal infection. All 8 MAb improved survival rates by 33%-100% against a high dose lethal challenge with mouse-adapted ZEBOV. This work has important implications for further development of vaccines and immunotherapies for ZEBOV infection.