Merck
  • Comparison between haplotype-based and individual snp-based genomic predictions for beef fatty acid profile in Nelore cattle.
Comparison between haplotype-based and individual snp-based genomic predictions for beef fatty acid profile in Nelore cattle.
Journal of animal breeding and genetics = Zeitschrift fur Tierzuchtung und Zuchtungsbiologie (2019-12-24)
Fabieli Loise Braga Feitosa, Angélica Simone Cravo Pereira, Sabrina Thaise Amorim, Elisa Peripolli, Rafael Medeiros de Oliveira Silva, Camila Urbano Braz, Adrielle Matias Ferrinho, Flavio Schramm Schenkel, Luiz Fernando Brito, Rafael Espigolan, Lucia Galvão de Albuquerque, Fernando Baldi
ABSTRACT
The aim of this study was to evaluate the genomic predictions using the single-step genomic best linear unbiased predictor (ssGBLUP) method based on SNPs and haplotype markers associated with beef fatty acids (FAs) profile in Nelore cattle. The data set contained records from 963 Nelore bulls finished in feedlot (±90 days) and slaughtered with approximately 24 months of age. Meat samples from the Longissimus dorsi muscle were taken for FAs profile measurement. FAs were quantified by gas chromatography using a SP-2560 capillary column. Animals were genotyped with the high-density SNP panel (BovineHD BeadChip assay) containing 777,962 markers. SNPs with a minor allele frequency and a call rate lower than 0.05 and 0.90, respectively, monomorphic, located on sex chromosomes, and with unknown position were removed from the data set. After genomic quality control, a total of 469,981 SNPs and 892 samples were available for subsequent analyses. Missing genotypes were imputed and phased using the FImpute software. Haplotype blocks were defined based on linkage disequilibrium using the Haploview software. The model to estimate variance components and genetic parameters and to predict the genomic values included the random genetic additive effects, fixed effects of the contemporary group and the age at slaughter as a linear covariate. Accuracies using the haplotype-based approach ranged from 0.07 to 0.31, and those SNP-based ranged from 0.06 to 0.33. Regression coefficients ranged from 0.07 to 0.74 and from 0.08 to 1.45 using the haplotype- and SNP-based approaches, respectively. Despite the low to moderate accuracies for the genomic values, it is possible to obtain genetic progress trough selection using genomic information based either on SNPs or haplotype markers. The SNP-based approach allows less biased genomic evaluations, and it is more feasible when taking into account the computational and operational cost underlying the haplotypes inference.
Sigma-Aldrich
Tricosanoic acid, ≥99% (capillary GC)