Merck
  • Detection of nickel and palladium contact hypersensitivity by a flow cytometric lymphocyte proliferation test.
Detection of nickel and palladium contact hypersensitivity by a flow cytometric lymphocyte proliferation test.
Allergy (2014-12-03)
I Spoerri, K Scherer, S Michel, S Link, A J Bircher, I A F M Heijnen
ABSTRACT
We established a flow cytometric lymphocyte proliferation test (LPT) for the detection of nickel (Ni) and palladium (Pd) sensitization. Eighty-one consecutive patients with an indication for patch test (PT) were tested by LPT with Ni (NiSO4 ) and Pd (Na2 PdCl4 and PdCl2 ) salts. The imprecision of the LPT was low (coefficient of variation 7.2%). Using PT as a diagnostic reference, the sensitivity and specificity of LPT were 74.4% and 80% for NiSO4 , 74.4% and 78.3% for Na2 PdCl4 , and 57.2% and 85.4% for PdCl2 , respectively. For both Ni and Pd, the likelihood ratio for a positive PT markedly increased with increasing LPT value. With medical history as a reference, the sensitivity and specificity were 40.6% and 82.1% for LPT and 59.4% and 89.7% for PT, respectively. Combination of LPT and PT resulted in a higher specificity of 95%, albeit lower sensitivity of 34.4%. In conclusion, flow cytometric LPT represents a reliable and useful method for the detection of Ni and Pd sensitization. LPT values correlate with PT results and, when used in combination with PT, increase test specificity.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, ≥95.0% (HPLC)
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride ChemBeads
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Palladium(II) chloride, Vetec, reagent grade, 98%