description
Oval tips, stainless steel, amber 15G, 0.50" 50/box
Application
Oval tips provide a consistent flat-ribbon deposit. Optimum oval tips are precisely manufactured to make high-flow, high-viscosity dispensing easier. Burr-free, polished, passivated stainless steel dispensing tips with polypropylene SafetyLok hubs for a secure fit to barrel reservoirs.
Choose from one of the most recent versions:
Certificates of Analysis (COA)