MilliporeSigma
Skip To
MPD42
Alkali Scientific™
Alkali Scientific™ 48-Well Deep Well Plate
4.6 mL, U-Bottom, Square well, non-5/pk QTY 50
Select a Size
Change View
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Alkali Scientific™
General description
Deep Well Plates U-BottomZoom Deep Well Plates U-Bottom * USP grade VI Polypropylene material; High chemical stability; Autoclaved by high temperature; For multi-channel pipette and automatic equipment * Conical bottom design to minimize the residual volume * Clear alphanumeric codes for identification * Conform to SBS/ ANSI * Stackable, reliable tight sealing * Non-pryogenic, DNase/RNase free * Both E-beam sterilized and non-sterilized deep well plates are available
Choose from one of the most recent versions:
Certificates of Analysis (COA)