Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
description
Cell line type: GPCR Cell Lines
GPCR Class A
protein target M1
Quality Level
technique(s)
calcium flux assay: suitable
detection method
fluorometric
Gene Information
human ... CHRM1(1128)
General description
Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.
The muscarinic acetylcholine receptor family consists of five GPCRs that mediate some of the neurotransmission functions of acetylcholine in the CNS and the periphery. The M1 receptor, along with the M3 and M5 receptors, signal through Gq/11 and subsequent release of Ca++ from the ER. The M1 receptor is expressed in ganglia and mediates depolarization of ganglia by inhibition of voltage-gated M-type K+ channels. In addition, the M1 receptor mediates venous contraction (Caulfield and Birdsall, 1998). Millipore′s cloned human M1 -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant M1 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at M1.
The muscarinic acetylcholine receptor family consists of five GPCRs that mediate some of the neurotransmission functions of acetylcholine in the CNS and the periphery. The M1 receptor, along with the M3 and M5 receptors, signal through Gq/11 and subsequent release of Ca++ from the ER. The M1 receptor is expressed in ganglia and mediates depolarization of ganglia by inhibition of voltage-gated M-type K+ channels. In addition, the M1 receptor mediates venous contraction (Caulfield and Birdsall, 1998). Millipore′s cloned human M1 -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant M1 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at M1.
Components
Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.
Disclaimer
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service