A9104
AKT3, active human
buffered aqueous glycerol solution, protein apparent mol wt ~83 kDa, ≥85% (SDS-PAGE), recombinant, expressed in E. coli
Synonym(s):
PKBγ, Protein Kinase B γ, RAC-gamma serine/threonine-protein kinase, STK-2
About This Item
recombinant
expressed in E. coli
assay
≥85% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
300-3000 units/mg protein
mol wt
protein apparent mol wt ~83 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... AKT3(10000)
Application
Biochem/physiol Actions
Unit Definition
Physical form
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service