Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3658

Sigma-Aldrich

Thioredoxin from Spirulina sp.

lyophilized powder, 1.5-6 A650 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

biological source

Spirulina

assay

≥70% (GE)

form

lyophilized powder

specific activity

1.5-6 A650 U/mg

composition

Protein, ~70% Lowry

technique(s)

activity assay: suitable

storage temp.

2-8°C

Gene Information

Spirulina ... trxA(41381942)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mediates the reduction of disulfide bonds in proteins.
Thioredoxin from Spirulina sp. was shown to inhibit the autophosphorylation of the S-locus receptor kinase (SRK).

Unit Definition

One unit will cause a ΔA650 of 1.0 in 1 min at 25 °C in the insulin reduction assay.

Physical form

Lyophilized powder containing potassium phosphate buffer

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Cabrillac et al.
Nature, 410(6825), 220-223 (2001-03-10)
The self-incompatibility response in Brassica allows recognition and rejection of self-pollen by the stigmatic papillae. The transmembrane S-locus receptor kinase (SRK), a member of the receptor-like kinase superfamily in plants, mediates recognition of self-pollen on the female side, whereas the
Effects of thioredoxin-1 on neurogenesis after brain ischemia/reperfusion injury.
Feng Zhou et al.
CNS neuroscience & therapeutics, 19(3), 204-205 (2013-02-28)
Philippe Lanotte et al.
PloS one, 8(1), e54393-e54393 (2013-02-02)
Group B streptococcus (GBS, Streptococcus agalactiae) is a leading cause of meningitis and sepsis in newborns and an etiological agent of meningitis, endocarditis, osteoarticular and soft tissue infections in adults. GBS isolates are routinely clustered in serotypes and in genotypes.
Jackelyn M Kembro et al.
Biophysical journal, 104(2), 332-343 (2013-02-28)
To understand the mechanisms involved in the control and regulation of mitochondrial reactive oxygen species (ROS) levels, a two-compartment computational mitochondrial energetic-redox (ME-R) model accounting for energetic, redox, and ROS metabolisms is presented. The ME-R model incorporates four main redox
Txnip, tanycytes, and torpor.
Francis J P Ebling et al.
Endocrinology, 154(6), 1970-1972 (2013-05-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service