storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
HyStem®-HP Hydrogel Kit - The growth factor delivery matrix HyStem-HP hydrogel is fully chemically-defined and is ideal for cell applications whereby the slow, continuous release of growth factors is crucial to re-creating a desired microenvironment. The HyStem-HP Hydrogel Kit contains a combination of thiol-modified hyaluronan and a thiol-modified heparin (Heprasil), thiol-modified denatured collagen (Gelin-S), and thiolreactive crosslinker, PEGDA (Extralink). HA is the simplest glycosaminoglycan (a negatively charged, linear, non-sulfated polysaccharide) that provides compression strength, lubrication, and hydration. HA regulates cell adhesion, motility, proliferation and differentiation. The immobilized heparin in the HyStem-HP hydrogel mimics the heparin sulfate proteoglycans normally present in the extracellular matrix. Heparin forms an ionic bond with proteins and protects them from proteolysis and facilitates their slow release into the cell culture medium. This significantly reduces the amount of growth factor required to trigger cell growth or differentiation compared to when growth factors are added directly to the medium. Product may be shipped ambient, but please store components at the temperature stated on the vials until ready for use.
Application
GS315F replaces Sigma HYSHP020 with a new user-friendly buffering sytem. Follow preparation instructions found in the product information sheet.
Features and Benefits
Features - Growth factors can be mixed into the hydrogels prior to gelation to provide a slow growth factor release depot.- - Hydrogels are suitable for animal implantation (such as angiogenesis applications and cell or drug delivery), culturing of primary cells, stem cells, and cell lines in the presence of growth factors. - Cells can be encapsulated or grown on the hydrogel surface in any format, including culture flasks, 6- to 384-well plates or tissue culture inserts. - Hydrogels can be easily customized by the user to possess the desired stiffness and gelation time by manipulating component concentration and mixing ratios.
Gelation Reconstituted HyStem-HP components remain liquid at 15 to 37°C. The hydrogel is formed when the crosslinking agent, Extralink® (PEGDA) is added to a mixture of Heprasil (thiol-modified hyaluronan plus heparin) and Gelin-S (thiol-modified gelatin). Gelation occurs in about twenty minutes after all three components are mixed. No steps depend on low temperatures or low pH. Diluting the components with phosphate-buffered saline (PBS) or cell-culture medium can increase the gelation time.
3D Cell Recovery Matrix For digesting the matrix and recovering cells, we recommend using a Collagenase/Hyaluronidase mixture.
Gelation Reconstituted HyStem-HP components remain liquid at 15 to 37°C. The hydrogel is formed when the crosslinking agent, Extralink® (PEGDA) is added to a mixture of Heprasil (thiol-modified hyaluronan plus heparin) and Gelin-S (thiol-modified gelatin). Gelation occurs in about twenty minutes after all three components are mixed. No steps depend on low temperatures or low pH. Diluting the components with phosphate-buffered saline (PBS) or cell-culture medium can increase the gelation time.
3D Cell Recovery Matrix For digesting the matrix and recovering cells, we recommend using a Collagenase/Hyaluronidase mixture.
Legal Information
Extralink is a registered trademark of Glycosan BioSystems, Inc.
Hystem is a registered trademark of BIOTIME, INC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service