Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody (DEAE)

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL11(6356)

General description

Eotaxin Antibody: Chemokines play a key role in inflammation. The CC chemokine eotaxin is a potent and specific eosinophil chemoattractant that is expressed by a variety of cell types in certain inflammatory conditions. Some G-protein coupled chemokine receptors are also utilized as virus coreceptors for fusion and infection of cells. The eotaxin receptor CCR3 is required for HIV-1 entry into target cells such as microglia and eotaxin inhibits the infection of HIV-1.

Immunogen

Eotaxin antibody was raised in rabbits against a peptide corresponding to amino acids near the carboxy terminus of human eotaxin.The immunogen is located within the last 50 amino acids of Eotaxin.

Application

Eotaxin antibody can be used for detection of eotaxin by Western blot at 1 μg/mL. The recombinant protein can be used as positive control and monomer and homodimer are detected at approximate 9 and 18 kDa. Antibody can also be used for immunocytochemistry starting at 5 μg/mL. For immunofluorescence start at 10 μg/mL.Antibody validated: Western Blot in human samples; Immunocytochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service