Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

6H6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IL3RA(3563)

General description

CD123 is the alpha chain of interleukin 3 receptor (IL-3R alpha). This subunit heterodimerizes with the interleukin 3 receptor beta chain (CD131), which is shared with other receptors. CD123 interacts with IL-3 specifically, but with low affinity, and association with the beta subunit confers high affinity binding to the receptor heterodimer. Both chains are required for signaling, but receptor activation and signal transduction depend on IL-3 binding to CD123 as the initial step.

Specificity

The mouse monoclonal antibody 6H6 recognizes an extracellular epitope of CD123 (interleukin 3 receptor alpha), a 60-70 kDa transmembrane protein expressed by myeloid precursors, megakaryocytes, macrophages, dendritic cells, mast cells, basophils, and some B cells. This antibody does not inhibit IL-3 binding to its receptor.

Immunogen

IL3 receptor alpha chain expressed on the surface of transiently transfected COS cells

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service