SAB5201768
Monoclonal Anti-Tcp1 Alpha - Rpe antibody produced in rat
clone 91a, purified immunoglobulin
Synonym(s):
Anti-AI528772, Anti-Ccta, Anti-Tric, Anti-p63
About This Item
biological source
rat
Quality Level
conjugate
phycoerythrin (R-PE) conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
91a, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 60 kDa
species reactivity
pig, guinea pig, Caenorhabditis elegans, monkey, Drosophila melanogaster, human, hamster, Saccharomyces cerevisiae, rat, mouse, rabbit, dog, bovine
concentration
1 mg/mL
technique(s)
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Tcp1(21454)
Related Categories
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service