Skip to Content
MilliporeSigma
All Photos(16)

Key Documents

ZRB1874

Sigma-Aldrich

Anti-PDK1 Antibody, clone 1E13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

3-phosphoinositide-dependent protein kinase 1 (UniProt: O15530; also known as EC:2.7.11.1, hPDK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E13, recombinant monoclonal

description

1E13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 63.15 kDa
observed mol wt ~59 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDPK1(5170)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects PDK1. It targets an epitope within 19 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding o 19 amino acids from the C-terminal half of human PDK1.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PDK1 in NIH3T3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PDK1 in Jurkat and PC12 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound recombinant Human PDK1 protein with a KD of 1.4 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): Serial dilutions from a representative lot detected PDK1 in ELISA application.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PDK1 in Jurkat cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PDK1 in human small intestine and human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PDK1 in NIH3T3 cell lysate.

Target description

3-phosphoinositide-dependent protein kinase 1 (UniProt: O15530; also known as EC:2.7.11.1, hPDK1) is encoded by the PDPK1 (also known as PDK1) gene (Gene ID: 5170) in human. PDK-1 is a serine/threonine protein kinase that serves as a master kinase, phosphorylating and activating a subgroup of the AGC family of protein kinases. Its protein kinase domain is localized near the N-terminal (aa 82-342) and the pleckstrin homology domain (aa 459-550) resides near the C-terminal region. Its targets include Akt, p70 and p90 ribosomal protein S6 kinases, PKA, PKC, PAK-1, and serum and glucocorticoid-inducible kinase. It plays a key role in the transduction of signals from insulin by providing the activating phosphorylation to Akt 1, thus propagating the signal to downstream targets controlling cell proliferation and survival, as well as glucose and amino acid uptake and storage. It is reported to negatively regulate the TGF-β- induced signaling by modulating the association of SMAD3 and SMAD7 with TGF-β receptor and preventing the nuclear translocation of SMAD3 and SMAD4 as well as the translocation of SMAD7 from the nucleus to the cytoplasm in response to TGF-β. Its homodimerization is known to regulate its activity by maintaining the kinase in an autoinhibitory conformation. PDK-1 activity can be downregulated by NPRL2 that interferes with its phosphorylation at tyrosine 9, 373, and 376. NPRL2 also down-regulates its activity by interfering with tyrosine phosphorylation at the tyrosine 9, 373, and 376. The 14-3-3 protein theta (YWHAQ) also acts as its negative regulator by association with the residues surrounding the serine 241. Phosphorylation on serine 241 in the activation loop is required for its full activity. PDK-1 itself can autophosphorylate serine 241, leading to its own activation. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Seong, HA., et al. (2007). J. Biol. Chem. 282(16); 12272-12289; Lim, MA., et al. (2003). Proc. Natl. Acad. Sci. USA. 100(24); 14006-14011; Alessi, DR., et al. (1997). Curr. Biol. 7(4); 261-269).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service