Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 124.96 kDa
observed mol wt ~250 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... ABL1(25)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3F7 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects BCR-ABL1 phosphorylated on tyrosine 245 of ABL1, isoform 2.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphotyrosine 245 of human ABL1, isoform 2.

Application

Quality Control Testing

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected BCR-ABL1 (pTyr245) in K562 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho BCR-ABL1 (Tyr245) in MCF-7 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound phospho-ABL1 (Tyr245) peptide with a KD of 8.4 x 10-9 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-ABL1 (Tyr245) in K562 cells.

Peptide Inhibition Assay: Target band detection in a lysate from NIH3T3 cells was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tyrosine-protein kinase ABL1 (UniProt: also known as EC:2.7.10.2, Abelson murine leukemia viral oncogene homolog 1, Abelson tyrosine-protein kinase 1, Proto-oncogene c-Abl, p150) is encoded by the ABL1 (also known as ABL, JTK7) gene (Gene ID: 25) in human. ABL1 is a non-receptor tyrosine protein kinase that plays a role in many KEY® processes linked to cell growth and survival. It is detected in the nucleus and cytoplasm and shuttles between them depending on environmental signals. It is sequestered into the cytoplasm through interaction with 14-3-3 proteins. In the nucleus it displays DNA-binding activity and is involved in DNA-damage response and apoptosis. It can also localize to mitochondria in response to oxidative stress. ABL1 phosphorylates multiple receptor tyrosine kinases and is reported to promote endocytosis of EGFR, facilitate the formation of neuromuscular synapses through MUSK, and inhibit PDGFRB-mediated chemotaxis. It is also involved in the endocytosis of activated B-cell receptor complexes. ABL1 also regulates multiple pathological signaling cascades during infection. Several known tyrosine-phosphorylated microbial proteins have been identified as ABL1 substrates. ABL1 is stabilized in the inactive form by an association between the SH3 domain and the SH2-TK linker region, interactions of the N-terminal cap, and contributions from an N-terminal myristoyl group and phospholipids. It is activated by autophosphorylation and by SRC-family kinase-mediated phosphorylation. Its phosphorylation at Tyr-70 by members of the SRC family of kinases disrupts SH3 domain-based autoinhibitory interactions and INTERMOLECULAR® associations and enhances its kinase activity. Phosphorylation at Tyr-226 and Tyr-393 also correlate with increased activity. BCR®-ABL1, a constitutively active form of ABL1 is known to cause Chronic myelogenous leukemia (CML) and phosphorylation of Tyr-245 on the ABL1 kinase domain is commonly used as a marker of BCR®-ABL1 activation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Luttman, JH., et al. (2021). Cell Commun. Signal. 19; Article 59; Nagar, B., et al. (2006). Mol. Cell. 21(6); 787-798; Nagar, B., et al. (2003). Cell. 112(6); 859-871).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
BCR is a registered trademark of European Commission
INTERMOLECULAR is a registered trademark of Intermolecular, Inc.
KEY is a registered trademark of Hanna Instruments
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service