Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C15H16ClNO2S·HCl
CAS Number:
Molecular Weight:
346.27
MDL number:
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

(S)-Methyl-2-(2-chlorophenyl)-2-((2-(thiophen-2-yl)ethyl)amino)acetate hydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

grade

certified reference material
TraceCERT®

Quality Level

agency

(Not traceable to any primary pharmacopeia standard)

API family

clopidogrel

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

ClC1=CC=CC=C1[C@@H](C(OC)=O)NCCC2=CC=CS2.Cl

InChI

1S/C15H16ClNO2S.ClH/c1-19-15(18)14(12-6-2-3-7-13(12)16)17-9-8-11-5-4-10-20-11;/h2-7,10,14,17H,8-9H2,1H3;1H/t14-;/m0./s1

InChI key

ZXANKCFSGFEBQW-UQKRIMTDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Other Notes

HCl form of Clopidogrel besilate EP Impurity F and Clopidogrel hydrochloride EP Impurity F

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service