Product Name
Mesaconic acid 99%
Product Number
131040
Product Brand
ALDRICH
CAS Number
498-24-8
Molecular Weight
130.10
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder |
TITRATION (T) NAOH 0.1M | 98.5 - 101.5 % |
PURITY (GC AREA %) | ≥ 98.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |