Product Name
Lauroyl chloride 98%
Product Number
156930
Product Brand
ALDRICH
CAS Number
112-16-3
Molecular Weight
218.76
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO LIGHT YELLOW OR ORANGE LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
AND/OR | |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5%-102.5%(WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 97.5% (MINIMUM) |
REVISED JUNE 7, 2006 BTR | |
ADDED HNMR AS AN ALTERNATE TEST. DELETED | |
WORD "STANDARD" FROM IR STATEMENT. | |
06/07/06 BTR |