Product Name
3,5-Dimethoxybenzoyl chloride 97%
Product Number
161713
Product Brand
ALDRICH
CAS Number
17213-57-9
Molecular Weight
200.62
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE TO GREY POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5-103.5% (WITH AGNO3) |
OR | |
TITRATION | 96.5-103.5% (WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED MARCH 28, 2007 RJM |