Product Name
Ethyl 2-oxocyclopentanecarboxylate 95%
Product Number
168092
Product Brand
ALDRICH
CAS Number
611-10-9
Molecular Weight
156.18
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW-GREEN LIQUID |
IDENTITY | CONFORMS TO STRUCTURE BY IR OR HNMR SPECTRA. |
GAS LIQUID | 94.0% (MINIMUM) |
REVISED FEBRUARY 18, 2008 RJM |