Product Name
1,1-Cyclohexanediacetic acid 98%
Product Number
171344
Product Brand
ALDRICH
CAS Number
4355-11-7
Molecular Weight
200.23
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder |
TITRATION (T) NAOH 0.1M | 97.5 - 102.5 % |
PURITY (GC AREA %) | ≥ 97.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |