Product Name
2-Acetylcyclopentanone 98%
Product Number
179779
Product Brand
ALDRICH
CAS Number
1670-46-8
Molecular Weight
126.15
Test | Specification |
---|---|
APPEARANCE | LIGHT YELLOW OR LIGHT ORANGE LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 428C OF EDITION I, | |
VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
TITRATION | 97.5% - 102.5% (WITH NAOH) |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED OCTOBER 29, 1996 JSB |