Product Name
4-Phenoxybenzaldehyde 98%
Product Number
211265
Product Brand
ALDRICH
CAS Number
67-36-7
Molecular Weight
198.22
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5%-102.5% (WITH HYDROXYLAMINE) |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED AUGUST 20, 2008 RJM |