Product Name
2,6-Dimethyl-4-heptanone 99%
Product Number
273848
Product Brand
ALDRICH
CAS Number
108-83-8
Molecular Weight
142.24
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED APRIL 11, 1994 JSB |