Product Name
9,10-Phenanthrenequinone 95%
Product Number
275034
Product Brand
ALDRICH
CAS Number
84-11-7
Molecular Weight
208.21
Test | Specification |
---|---|
APPEARANCE (COLOR) | Orange |
APPEARANCE (FORM) | Powder |
PURITY (HPLC AREA %) | ≥ 94.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |