Product Name
6-Fluoroindole 98%
Product Number
349593
Product Brand
ALDRICH
CAS Number
399-51-9
Molecular Weight
135.14
Test | Specification |
---|---|
APPEARANCE | WHITE TO PINK POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 97.5% (MINIMUM) |
REVISED MARCH 7, 2007 JLH |