Product Name
(+)-Dihydrocarvone mixture of isomers
Product Number
37275
Product Brand
ALDRICH
CAS Number
5524-05-0
Molecular Weight
152.23
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow and Colorless to Light Green-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 95.0 % (SUM OF ISOMERS) |
SPECIFIC ROTATION (20/D) | 18 - 22 Degrees |
CONCENTRATION | NEAT |
REFRACTIVE INDEX N20/D | 1.470 - 1.472 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |