Product Name
Vanadium(V) oxytriisopropoxide
Product Number
404926
Product Brand
ALDRICH
CAS Number
5588-84-1
Molecular Weight
244.20
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW-GREEN TO GOLD OR ORANGE |
LIQUID | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 20.3%-21.4%V (WITH KMNO4) |
APPROVED JANUARY 20, 2005 RJM |