Product Name
4,N,N-Trimethylaniline ≥98.5% (GC)
Product Number
41690
Product Brand
ALDRICH
CAS Number
99-97-8
Molecular Weight
135.21
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Dark Green-Yellow to Dark Brown-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 98.5 % |
DENSITY D20/4 | 0.930 - 0.945 |
REFRACTIVE INDEX N20/D | 1.546 - 1.548 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 10 MONTHS |