Product Name
Cinchonine monohydrochloride hydrate 99%
Product Number
420328
Product Brand
ALDRICH
CAS Number
206986-88-1
Molecular Weight
330.85 (anhydrous basis)
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER, CRYSTALS, CRYSTALLINE POWDER |
AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.0%-101.0% (DRIED BASIS) (WITH AGNO3) |
OPTICAL ROTATION | +101(+/-2) DEGREES (C=1%, CHCL3) |
HIGH PRESSURE LIQUID | 98.5% (MINIMUM) |
LOSS ON DRYING | 2.0% LOSS (MINIMUM) |
OR | |
TITRATION | 2.0% H2O (MINIMUM) (WITH "KARL FISCHER" |
REAGENT) | |
REVISED OCTOBER 24, 2007 RJM |