Product Name
2-Ethynylanisole 97%
Product Number
467227
Product Brand
ALDRICH
CAS Number
767-91-9
Molecular Weight
132.16
Test | Specification |
---|---|
Appearance (Color) | Colorless to Brown |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Proton NMR spectrum | Conforms to Structure |
Purity (GC) | ≥96.5 % |