Product Name
Glycolic acid nitrile solution ~70% in H2O
Product Number
50640
Product Brand
ALDRICH
CAS Number
107-16-4
Molecular Weight
57.05
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
TITRATION | 69 - 71 % |
TITRATION (METHOD) | N-DETERMINATION ACCORDING TO KJELDAHL |
REFRACTIVE INDEX N20/D | 1.388 - 1.390 |
PH-TEST | ≤ 3.0 |
RECOMMENDED RETEST PERIOD | 42 MONTHS |