Product Name
4-Methoxybenzenesulfonamide 97%
Product Number
544655
Product Brand
ALDRICH
CAS Number
1129-26-6
Molecular Weight
187.22
Test | Specification |
---|---|
Appearance (Color) | White to Off-White |
Appearance (Form) | Conforms to Requirement |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Proton NMR spectra | Conforms to Structure |
Carbon | 43.3 - 46.5 % |
Nitrogen | 7.2 - 7.7 % |