Product Name
Quinacridonequinone
Product Number
634484
Product Brand
ALDRICH
CAS Number
1503-48-6
Molecular Weight
342.30
Test | Specification |
---|---|
APPEARANCE | YELLOW TO ORANGE POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 65.3%-78.6% |
NITROGEN 7.2%- 9.2% | |
APPROVED JULY 12, 2007 RJM |