Product Name
Chloro(η2-P,C-tris(2,4-di-tert-butylphenyl)phosphite)(tricyclohexylphosphine)palladium(II)
Product Number
667315
Product Brand
ALDRICH
CAS Number
502964-53-6
Molecular Weight
1068.22
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
IDENTITY | CONFORMS TO STRUCTURE BY IR, HNMR OR PNMR. |
ELEMENTAL ANALYSIS | CARBON 64.7 - 70.2% |
ICP ASSAY | CONFIRMS PHOSPHORUS AND PALLADIUM COMPONENTS |
APPROVED DECEMBER 18, 2006 LXG |