Product Name
4-(4-Fluorophenyl)-1H-imidazole 97%
Product Number
687065
Product Brand
ALDRICH
CAS Number
65020-70-4
Molecular Weight
162.16
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN SOLID, POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 64.3% TO 69.0% |
NITROGEN 16.7% TO 17.9% | |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED JULY 31, 2007 DJP |