Product Name
1,3-Diiodo-5,5-dimethylhydantoin ≥96%
Product Number
711624
Product Brand
ALDRICH
CAS Number
2232-12-4
Molecular Weight
379.92
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow and Faint Brown to Light Brown |
APPEARANCE (FORM) | Powder |
TITRATION | 96.0 - 104.0 % |
TITRATION (METHOD) | ARGENTOMETRIC AFTER OXYGEN COMBUSTION |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 36 MONTHS |