Product Name
1,3-Adamantanediol ≥99.0% (GC)
Product Number
93749
Product Brand
ALDRICH
CAS Number
5001-18-3
Molecular Weight
168.23
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 99.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |