Product Name
4-Bromothiophenol 95%
Product Number
B81956
Product Brand
ALDRICH
CAS Number
106-53-6
Molecular Weight
189.07
Test | Specification |
---|---|
Appearance (Color) | White to Yellow |
Appearance (Form) | Conforms to Requirement |
Crystals | |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 94.0 - 106.0 % |
Purity (GC) | ≥94.0 % |