Product Name
Diglycolic anhydride technical grade, 90%
Product Number
D103705
Product Brand
ALDRICH
CAS Number
4480-83-5
Molecular Weight
116.07
Test | Specification |
---|---|
APPEARANCE | WHITE CRYSTALS, CRYSTALLINE POWDER AND/OR |
CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 88.5% (MINIMUM) (WITH MORPHOLINE) |
REVISED JULY 23, 2003 JSB |