Product Name
2,4-Diamino-6-phenyl-1,3,5-triazine 97%
Product Number
D23408
Product Brand
ALDRICH
CAS Number
91-76-9
Molecular Weight
187.20
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER OR GRANULAR POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 55.7% - 59.8% |
NITROGEN 36.1% - 38.7% | |
OR | |
TITRATION | 96.5% - 103.5% (WITH HCLO4) |
HIGH PRESSURE LIQUID | 96.5% (MINIMUM) |
REVISED JUNE 6, 2008 BTR |