Product Name
4-Nitrobenzyl bromide 99%
Product Number
N13054
Product Brand
ALDRICH
CAS Number
100-11-8
Molecular Weight
216.03
Test | Specification |
---|---|
APPEARANCE | WHITE TO YELLOW TO TAN POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.5-101.5% (WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED APRIL 15, 2008 RJM |